Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ionic Strength Adjustor, pH Indicating for use with Ammonia Selective Electrode, Reagents

Catalog No. 01334908
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-908 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-908 Supplier Reagents Holdings Llc Supplier No. CI2804001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ionic Strength Adjustor, pH Indicating for use with Ammonia Selective Electrode, Reagent, use for Ammonia analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Packaging Poly Bottle
Specific Gravity 1.1205
Physical Form Liquid
Color Dark Blue
pH 14
Boiling Point 103°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Chemical Name or Material Ionic Strength Adjustor, pH Indicating for use with Ammonia Selective Electrode
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.