Learn More
Pyramex™ Ionix™ Safety Eyewear
Catalog No. 19155834 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Blue, Mirror
Clear
Gray
Silver, Mirror
Lens Coating:
UV-treated
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating
19-155-834 Gray -
19-155-831 Clear -
19-155-832 Clear UV-treated
19-155-835 Gray UV-treated
19-155-836 Blue, Mirror -
19-155-837 Silver, Mirror -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-155-834 Supplier Pyramex™ Supplier No. SB8120D
This item is not returnable. View return policy
Extreme style in safety eyewear, with clean lines and sports appeal
  • Extreme style in safety eyewear
  • Built-in rubber nose piece
  • Ventilated, straight-back flexible temples
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Passes MIL-PRF 32432 High Velocity Impact Standards.
  • Specifications
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Lens Tint Gray
    Frame Color Black
    Weight (Metric) 26 g
    Lens Material Polycarbonate
    Frame Material Nylon