Learn More
TARGETMOL CHEMICALS INC Ionomycin 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T72851MG
Encompass_Preferred
Ionomycin is an calcium ionophore and an antibiotic that binds calcium ions (Ca2+). It is produced by the bacterium Streptomyces conglobatus.It is used in research to raise the intracellular calcium level (Ca2+) and as a research tool to understand Ca2+ transport across biological membranes. Ionomycin promotes apoptosis and induces the activation of protein kinase C (PKC). Purity 99.54%
Catalog No. 50-287-8208
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.