Learn More
Apexbio Technology LLC Ionomycin calcium salt 56092-82-1 10mg
Supplier: Apexbio Technology LLC B516510
Ionomycin calcium salt is a calcium ionophore produced by Streptomyces species that facilitates rapid transportation of calcium ions across biological membranes Within cellular environments ionomycin induces increased intracellular Ca concentration triggering downstream signaling pathways and cytokine release including interleukin (IL)-2 IL-4 perforin and interferon production In reproductive biology studies ionomycin is routinely utilized to initiate oocyte activation regulate embryonic developmental processes and investigate calcium-dependent fertilization mechanisms It is widely applied as a research tool in immunology assays requiring elevation of intracellular calcium levels The IC50 for ionomycin-mediated calcium transport typically ranges around 10 mol/L in cell-based assays Currently ionomycin remains limited to in vitro and preclinical research contexts
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.