Learn More
Abcam Ionomycin Free Acid, Ca2+ Ionophore, Ethanol Solution, 5 mg
Description
- Molecular Weight: 709 Da
- Purity: >97%
- Form: Liquid
- Potent and highly selective Ca²± ionophore: Effective mobile ion carrier for Ca²± and highly specific for divalent cations (Ca > Mg >> Sr = Ba)
- Applications: Useful for studies of Ca²± transport across biological membranes and increasing intracellular Ca²± concentrations
- Complexation: Complexes with Ca²± between pH 7 and 9.5
- Absorption Maximum: 280 nm (MeOH): 18,600 M-1cm-1
- Storage Buffer: 98.73% Ethanol, 1.27% Ionomycin free acid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Calcium |
| Substrate Properties | Chemical Substrate |
| Substrate | Ca2+ |
| Shipping Condition | Ambient |
| Form | Liquid |
| Content And Storage | Store at -20°C. |
| Buffer | 98.73% ethanol, 1.27% ionomycin free acid |
| Cross Reactivity | Ca2+, Mg2+, Sr2+, Ba2+ |
| Formulation | 98.73% Ethanol, 1.27% Ionomycin free acid |
| Quantity | 5 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.