Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iopanoic Acid 98.0+%, TCI America™
SDP

Catalog No. I030025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I030025G 25 g
I03005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I030025G Supplier TCI America Supplier No. I030025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96-83-3
Molecular Formula C11H12I3NO2
Molecular Weight (g/mol) 570.935
MDL Number MFCD00038687
InChI Key OIRFJRBSRORBCM-UHFFFAOYSA-N
Synonym 3-Amino-alpha-ethyl-2,4,6-triiodohydrocinnamic Acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropionic Acid, Iodopanoic Acid, 2-(3-Amino-2,4,6-triiodobenzyl)butyric Acid
PubChem CID 3735
ChEBI CHEBI:5951
IUPAC Name 2-[(3-amino-2,4,6-triiodophenyl)methyl]butanoic acid
SMILES CCC(CC1=C(C(=C(C=C1I)I)N)I)C(=O)O

Specifications

Melting Point 153°C
Quantity 25 g
Formula Weight 570.94
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Iopanoic Acid
EINECSNumber (9)-0171&(3)-3088
RTECSNumber MW5075000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.