Learn More
TARGETMOL CHEMICALS INC IOPROMIDE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1522
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Iopromide (Iopromidum) is a low osmolar non-ionic contrast agent for intravascular use. purity: 99%
Catalog No. 50-258-1242
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.