Learn More
TARGETMOL CHEMICALS INC L-Octanoylcarnitine 10MG
Supplier: TARGETMOL CHEMICALS INC T1939810MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.L-Octanoylcarnitine is the active octanoylcarnitine present in plasma and is a potential biomarker for breast cancer. Purity 99.63%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.