Learn More
Cayman Chemical IOX1, 5852-78-8, 5 mg
SDP
Supplier:  Cayman Chemical 115725
Encompass_Preferred
A broad-spectrum inhibitor of 2OG oxygenases that inhibits JMJD2 demethylase activity (IC50 = 87 μM); inhibits JMJD2A, JMJD2E and the 2OG oxygenases PHF8, PHD2, and FIH (IC50s = 1.7, 2.4, 13.3, 14.3, and 20.5 μM, respectively)
Catalog No. NC0393475
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.