Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IP-10/CXCL10 Matched Antibody Pair

Catalog No. 5018293
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-182-93 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-182-93 Supplier Invitrogen™ Supplier No. BMS6018MST
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Mouse IP-10 Matched Antibody Pair Kit comes with pre-matched antibody pairs, standards, sample dilutent and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IP-10. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL10 (IP-10)
Applications ELISA
Classification Polyclonal
Conjugate Biotin
Gene Accession No. P17515
Gene Alias Interferon-gamma-inducible protein 10, CXCL10
Gene Symbols Cxcl10
Quantity 10 x 96 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15945
Target Species Mouse
Content And Storage 1 vial (0.275 mL) polyclonal Coating Antibody to mouse IP-10 (100 ug/mL)
1 vial (55 uL) Biotin-Conjugate anti-mouse IP-10 polyclonal antibody
1 vial (22 uL) Streptavidin-HRP
10 vials mouse IP-10 Standard protein lyophilized, 1 ng/mL upon reconstitution
2 vials (50 mL) Sample Diluent
Product Type ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.