Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IP3 Receptor 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670201 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-201 Supplier Invitrogen™ Supplier No. PA3901A
  • $552.08
    $428.65 / Each of 1

    Save $123.43

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA3-901A detects inositol 1,4,5-triphosphate receptor type-I (IP3R-I) from mouse, human and rat tissues. PA3-901A has been successfully used in Western blot procedures. By Western blot, this antibody detects a ~240-260 kDa protein representing IP3R-I from rat and mouse brain extract. PA3-901A is a remanufactured lot of PA3-901 which has previously been used in Western blot, immunoprecipitation and immunohistochemistry procedures. The PA3-901A immunogen is a synthetic peptide corresponding to residues N(1829) K K K D D E V D R D A P S R K K A K E(1848) of human IP3 Receptor Type I. This sequence is 100% conserved in human, mouse, and rat IP3R-I protein. This peptide (Cat. # PEP-019) is available for use in neutralization and control experiments.

Intracellular channel that mediates calcium release from the endoplasmic reticulum following stimulation by inositol 1,4,5-trisphosphate. Involved in the regulation of epithelial secretion of electrolytes and fluid through the interaction with AHCYL1. Plays a role in ER stress-induced apoptosis. Cytoplasmic calcium released from the ER triggers apoptosis by the activation of CaM kinase II, eventually leading to the activation of downstream apoptosis pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP3 Receptor 1
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum, PBS with 0.05% sodium azide
Gene Itpr1
Gene Accession No. P11881, P29994, Q14643
Gene Alias ACV; CLA4; D6Pas2; ENSMUSG00000072853; Gm10429; I145TR; inositol 1,4,5-triphosphate receptor 1; inositol 1,4,5-triphosphate receptor, type 1; inositol 1,4,5-trisphosphate receptor 1; inositol 1,4,5-trisphosphate receptor type 1; inositol 1,4,5-trisphosphate receptor, type 1; inositol 1,4,5-trisphosphate-binding protein P400; InsP>3>R type I; Insp3r; InsP3R type I; insP3R1; IP3 receptor; IP3 receptor isoform 1; Ip3r; IP-3-R; IP3R 1; IP3R1; Itpr1; Itpr-1; opisthotonus; opt; P400; Pcd6; Pcp1; Pcp-1; PPP1R94; protein PCD-6; protein phosphatase 1, regulatory subunit 94; Purkinje cell protein 1; SCA15; SCA16; SCA29; SI-SIII-SIIA; Type 1 inositol 1,4,5-trisphosphate receptor; type 1 InsP3 receptor
Gene Symbols Itpr1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues N(1829) K K K D D E V D R D A P S R K K A K E(1848) of human IP3 Receptor Type I.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16438, 25262, 3708
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.