Learn More
Invitrogen™ IP3 Receptor 2 Polyclonal Antibody

Description
IP3 Receptor 2 Polyclonal Antibody for DB, Flow, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | IP3 Receptor 2 |
| Applications | Dot Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | ITPR2 |
| Gene Accession No. | P29995, Q14571, Q9Z329 |
| Gene Alias | AI649341; ANHD; CFAP48; cilia and flagella associated protein 48; inositol 1,4,5-triphosphate receptor 2; inositol 1,4,5-triphosphate receptor 5; inositol 1,4,5-triphosphate receptor, type 2; inositol 1,4,5-trisphosphate receptor type 2; inositol 1,4,5-trisphosphate receptor, type 2; inositol 1,4,5-trisphosphate type V receptor; inositol triphosphate receptor type 2; inositol trisphosphate receptor type 2; insP3R2; InsP3R-2; InsP3R-5; IP3 receptor; IP3 receptor isoform 2; IP3 receptor/Ca2+ channel type 2; IP3R; IP3R 2; IP3R2; Itpr2; Itpr5; LOW QUALITY PROTEIN: inositol 1,4,5-trisphosphate receptor type 2; si:dkey-196d8.1; Type 2 inositol 1,4,5-trisphosphate receptor; type 2 InsP3 receptor |
| Gene Symbols | ITPR2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.