Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IP3 Receptor 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1904 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1904 Supplier Invitrogen™ Supplier No. PA1904
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-904 detects inositol 1,4,5-trisphosphate receptor type-II (IP3R-II) from rat and mouse tissues. PA1-904 has been successfully used in immunohistochemistry, immunofluorescence and immunocytochemistry procedures. Immunohistochemical staining of IP3R-II in rat brain with PA1-904 results in staining of the hippocampus, corpus collosum, and cerebellum. This product fails to detect IP3R-II in Western blot procedures. PA1-904 immunizing peptide corresponds to amino acid residues 317-334 from rat IP3R-II. PA1-904 immunizing peptide (Cat. # PEP-024) is available for use in neutralization and control experiments.

Inositol 1,4,5-trisphosphate (IP3) is a second messenger for many growth factors, hormones, and neuro-transmitters. Upon binding to IP3R, IP3 triggers the release of intracellular, luminal calcium to the cytosol. Functional IP3R is a homo- or heterotetramer of ~240 kDa glycoprotein subunits. IP3R protein is structurally and functionally related to one other important intracellular calcium-release channel, the ryanodine receptor. The similarity between the two receptors continues at the physiological level owing to a physical association each receptor can have with the immunophilin protein, FKBP12. Phosphorylation of the IP3R by PKC causes an increase in IP3-mediated calcium release. Concomitantly, the phosphatase activity of calcineurin is stimulated upon its association with the FKBP12-IP3R complex. Calcium release is reduced when the PKC target site on the IP3R is dephosphorylated by calcineurin resulting in calcium oscillations. Mammalian IP3R subunits are the product of three distinct genes that are widely expressed and differentially regulated. IP3R type I (IP3R-I) has been detected in heart, liver, kidney, ovary, and Purkinje neurons of the cerebellum. IP3R-II is found predominantly in the brain. IP3R-III is known to be expressed in pancreatic islets, kidney, and the gastrointestinal tract.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP3 Receptor 2
Applications Immunohistochemistry (Frozen), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene ITPR2
Gene Accession No. P29995, Q14571, Q9Z329
Gene Alias AI649341; ANHD; CFAP48; cilia and flagella associated protein 48; inositol 1,4,5-triphosphate receptor 2; inositol 1,4,5-triphosphate receptor 5; inositol 1,4,5-triphosphate receptor, type 2; inositol 1,4,5-trisphosphate receptor type 2; inositol 1,4,5-trisphosphate receptor, type 2; inositol 1,4,5-trisphosphate type V receptor; inositol triphosphate receptor type 2; inositol trisphosphate receptor type 2; insP3R2; InsP3R-2; InsP3R-5; IP3 receptor; IP3 receptor isoform 2; IP3 receptor/Ca2+ channel type 2; IP3R; IP3R 2; IP3R2; Itpr2; Itpr5; LOW QUALITY PROTEIN: inositol 1,4,5-trisphosphate receptor type 2; si:dkey-196d8.1; Type 2 inositol 1,4,5-trisphosphate receptor; type 2 InsP3 receptor
Gene Symbols ITPR2
Host Species Rabbit
Immunogen Synthetic Peptide: E(317) L N P D Y R D A Q N E G K T V R D(334).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16439, 3709, 81678
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.