Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP6K2 Monoclonal Antibody (OTI1F12), TrueMAB™, OriGene
SDP

Catalog No. 501682386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2386 Supplier OriGene Supplier No. TA808811
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Inositol 1,4,5-trisphosphate (IP3) is a second messenger for many growth factors, hormones, and neurotransmitters and, upon binding to the IP3 receptor (IP3R), triggers the release of intracellular, luminal calcium to the cytosol. Several additional cellular forms of inositol phosphates have been discovered. These include some which have been recently discovered and characterized to possess up to seven or eight phosphate groups, including pyrophosphates (PP-IPs: pyrophosphate-inositol phosphates). It has been proposed that high-energy pyrophosphates may participate in protein phosphorylation. Inositol-hexakisphosphate (IP6) kinase, or PiUS (Pi Uptake Stimulator) has ATP synthase activity and transfers a phosphate from PP-IP5 to ADP to form ATP thus suggesting an important physiological role for PP-IP5 and IP6 kinase.

Specifications

Antigen IP6K2
Applications Western Blot
Classification Monoclonal
Clone OTI1F12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene IP6K2
Gene Accession No. Q9UHH9
Gene Alias IHPK2, PIUS
Gene Symbols IP6K2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-250 of human IP6K2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51447
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.