Learn More
Description
MA1-10886 detects PSE1/Ku Protein from human samples. MA1-10886 has been successfully used in Western blot applications. The MA1-10886 immunogen is Proximal Sequence Element (PSE1) of the human U1 small nuclear RNA extracted from human placenta.
Specifications
Specifications
| Antigen | IPO5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | N9C1 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | IPO5 |
| Gene Accession No. | O00410 |
| Gene Alias | 1110011C18Rik; 5730478E03Rik; AA409333; C76941; IMB3; imp5; importin 5; importin beta-3 subunit; importin subunit beta-3; Importin-5; IPO 5; IPO5; karyopherin (importin) beta 3; Karyopherin beta 3; karyopherin beta-3; Kpnb3; MGC2068; Pse1; RAN binding protein 5; Ran_GTP binding protein 5; ran-binding protein 5; RanBP 5; Ranbp5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
