Learn More
Cellular Engineering Technologies iPS Cell Growth Media KitComponents:iPS Base Media 500mLiPS Growth Supplement 7mL
Supplier:  Cellular Engineering Technologies MR1001K
Encompass_Preferred
This growth media kit is a universal, defined media for iPS cell systems formulated and validated to grow on Vitronectin XF, Geltrex, and Matrigel extracellular matrices. Media is serum-free and xeno-free and has been uniquely formulated to minimize cell differentiation, promote robust growth, and prevent cell death. This complete media does not require additional additives or reagents. Base media is shipped with gel packs. A separate growth supplement is shipped with dry ice.Components:iPS Base Media 500mLiPS Growth Supplement 7mL
Catalog No. NC2753058
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.