Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ipsapirone
GREENER_CHOICE

Catalog No. p-4784041
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-691-0 10 mg
18-695-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-691-0 Supplier Tocris Bioscience™ Supplier No. 1869/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective 5-HT1A agonist

Selective 5-HT1A receptor agonist (Ki = 10 nM). Anxiolytic in vivo.

Chemical Identifiers

CAS 95847-70-4
Molecular Formula C19H23N5O3S

Specifications

Quantity 10 mg
Formula Weight 401.48
Purity >99%
Molecular Weight (g/mol) 401.485
Chemical Name or Material Ipsapirone
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.