Learn More
Ubpbio IPTG, 100G
Supplier:  Ubpbio P1010100
Encompass_Preferred
Protein Purification Reagents – IPTG (UBPBio catalog# P1010-10, P1010-25, P1010-100) – IPTG, Isopropyl β-D-1-thiogalactopyranoside, binds the lac repressor and induces expression of β-galactosidase in E. coli. IPTG is widely used to induce the expression of cloned genes under the control of the lac operon and to screen blue/white colony in X-gal plate.
Catalog No. 50-112-6936
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.