Learn More
Carbosynth LLC. Isopropyl b-D-thiogalactopyranoside, 367-93-1, MFCD00063273, 250 g
SDP
Supplier:  Carbosynth LLC. EI05931250G
Encompass_Preferred
Chemical Formula: C9H18O5S, MW: 238.30, Synonyms: IPTG non-animal origin; IPTG. A non-metabolizable allolactose analogue, widely used in molecular biology for overexpression of recombinant proteins from inducible systems under the control of lac promoter.
Catalog No. NC1105120
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.