Learn More
Ubpbio 1M IPTG prepared in water and sterilized by filtration
Supplier:  Ubpbio P101110
Encompass_Preferred
IPTG, Isopropyl β-D-1-thiogalactopyranoside, binds the lac repressor and induces expression of β-galactosidase in E. coli. IPTG is widely used to induce the expression of cloned genes under the control of the lac operon and to screen blue/white colony in X-gal plate. Note: IPTG is often prepared in distilled water in 1 M stock solution (238 mg/ml). Sterile the solution using 0.2 um filter. Aliquot the stock and store under -20°C.
Catalog No. 50-233-9336
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.