Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Analytics™ IQ Express Docking Stations

Catalog No. 191501201
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
ToxiPro/LTD
Components Measured:
Oxygen, Carbon Monoxide, Hydrogen, Sulfur, Sulfur Dioxide, Chlorine, Nitrogen Dioxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Components Measured
19-150-1201 ToxiPro/LTD Oxygen, Carbon Monoxide, Hydrogen, Sulfur, Sulfur Dioxide, Chlorine, Nitrogen Dioxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-150-1201 Supplier Honeywell Analytics™ Supplier No. 54469100N

Please call customer service for current pricing. 1-800-766-7000

missing translation for 'validMainframeMsg'

Includes

Software, installation disks, and manuals.

Requires

Appropriate calibration gas for bump tests and span calibrations.

Automatic calibration station and data download center compatible with Sperian gas detectors

Just drop the detector into the dock, and the IQ Express does the rest—in as little as 20 seconds.

  • Identifies instrument and performs a battery check before moving on to bump test
  • Performs full span calibration if bump test fails; no user intervention is required
  • Tests audible, visible, and vibrating alarms
  • Downloads instrument data
  • Displays the word “Ready” when all tests and functions are completed

Order Info

Standard stations are suitable for detectors outfitted with O2, LEL, CO, or H2S sensors. Enhanced versions are suitable for those sensors, as well as SO2, Cl2, NO2, NH3, HCN and PH3.

TRUSTED_SUSTAINABILITY

Specifications

Components Measured Oxygen, Carbon Monoxide, Hydrogen, Sulfur, Sulfur Dioxide, Chlorine, Nitrogen Dioxide
For Use With (Equipment) ToxiPro/LTD
Type Docking Station
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.