Learn More
Beckman Coulter IQ OQ PROTOCAL REV A PAT700
Catalog No. NC3788502
Encompass
  • Provides documentation and procedures to meet global pharmacopeial requirements.
  • Ensures the instrument is properly installed and validated for reliable performance.
  • Verifies critical operating parameters and functions, ensuring consistent and accurate operation.
  • Offers a structured approach to qualification activities and documents results.
  • Critical for installation qualification and operational qualification of the instrument.
  • Structured documentation for compliance with USP <1058> standards.
  • Includes detailed checks for various firmware versions of supported instruments.
  • Facilitates adherence to regulatory standards for laboratory instruments.
Catalog No. NC3788502 Supplier Beckman Coulter Supplier No. D07176
Add to Cart
Add to Cart
Request a Quote
  • Provides documentation and procedures to meet global pharmacopeial requirements.
  • Ensures the instrument is properly installed and validated for reliable performance.
  • Verifies critical operating parameters and functions, ensuring consistent and accurate operation.
  • Offers a structured approach to qualification activities and documents results.
  • Critical for installation qualification and operational qualification of the instrument.
  • Structured documentation for compliance with USP <1058> standards.
  • Includes detailed checks for various firmware versions of supported instruments.
  • Facilitates adherence to regulatory standards for laboratory instruments.