Learn More
Bulwark™ FR IQ Series™ Long Sleeve Comfort Woven Lightweight Shirt Tall
Shop All Red Kap Industries ProductsDescription
- Fabric: Bulwark Exclusive 5.5 oz. 50% Aramid 25% Lyocell 25% Modacrylic- Flame-resistant
- Care: Industrial Laundry - Light Soil, Home Wash
- Banded, Topstitched collar with red trim at neck seam
- Topstitched cuffs with two-button adjustable closures
- Tailored sleeve placket
- Hemmed front placket with button closure
- Larger patch chest pockets
- Full side seam gusset
- iQ Series™ seam tag
- iQ Series™ hang tag
- Polybag
- Hanger with size clip
- Protection: Arc Rating ATPV 8.5 calories/cm2;
- Country of Origin: Imported
Specifications
Specifications
| Size | 5X-Large |
| Gender | Men's |
| Material | Aramid, Lyocell, Modacrylic |
| Color | Navy |
| Neck Style | Collar |
| Closure Type | Button |
| No. of Pockets | 2 |
| Arc Flash Rating | ATPV 8.5 calories/cm2 |
| Certifications/Compliance | NFPA 2112, Standard on Flame- Resistant Garments for Protection of Industrial Personnel Against Flash Fire, 2018 Edition, EN ISO 11612:2015, Protective clothing against heat and flame; EN 340:2003, Protective clothing - General requirements; IEC 61482- 2:2009, Live working - Protective clothing against the thermal hazards of an electric arc |
| Size (English) Chest | 62.5 to 66 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.