Learn More
Sartorius iQue Annexin V 5x384
Supplier:  Sartorius 91030
Encompass
Just add and read for fast easily automated workflowMultiplex with Qbeads PlexScreen kits for richer content.PrincipleSurface expression of phosphatidylserine is detected by the binding of Annexin-V to the cell surface.
Catalog No. 50-289-1602