Learn More
Sartorius iQue Caspase 3/7 5x384
Supplier:  Sartorius 91034
Encompass
No wash protocol – just add and read for fast easily automated workflow.Multiplex with Qbeads PlexScreen kits for richer content.PrincipleThe activation of Caspase 3 and 7 is detected by the use of NucView 488 Caspase-3/7 substrate which upon cleavage by activated enzyme results in an intracellular fluorescent signal.
Catalog No. 50-289-1662