Learn More
Sartorius iQue Human 4-Plex Apoptosis
Catalog No. 502891591 Shop All Sartorius Products
Encompass
Look no further for quick accurate and reliable apoptosis assay determination – the iQue Human 4-Plex Apoptosis Kit ensures top-rate determinations and easily replicable results. We make it easy for you to obtain robust data. Simple Multiparameter Analysis With the iQue Human 4-Plex Apoptosis Kit perform simple no-wash multiparameter assays designed for measuring multiple cellular endpoints. This kit was designed to be easy to use accurate and provide roust and distinct information in apoptosis assays. Easy Streamlined Workflow iQue Human 4-Plex Apoptosis Kit allows for comprehensive apoptosis analysis with unparalleled ease. Like many other iQue Cell-Based Kits the iQue Human 4-Plex Apoptosis Kit has been optimized for no-wash protocol. You need only add and read for a fast easily streamlined workflow. Perform 4-plex analysis with greater ease than many single endpoint kits.
Catalog No. 50-289-1591 Supplier Sartorius Supplier No. 90053
Add to Cart
Edge
Add to Cart
Look no further for quick accurate and reliable apoptosis assay determination – the iQue Human 4-Plex Apoptosis Kit ensures top-rate determinations and easily replicable results. We make it easy for you to obtain robust data. Simple Multiparameter Analysis With the iQue Human 4-Plex Apoptosis Kit perform simple no-wash multiparameter assays designed for measuring multiple cellular endpoints. This kit was designed to be easy to use accurate and provide roust and distinct information in apoptosis assays. Easy Streamlined Workflow iQue Human 4-Plex Apoptosis Kit allows for comprehensive apoptosis analysis with unparalleled ease. Like many other iQue Cell-Based Kits the iQue Human 4-Plex Apoptosis Kit has been optimized for no-wash protocol. You need only add and read for a fast easily streamlined workflow. Perform 4-plex analysis with greater ease than many single endpoint kits.