Learn More
Sartorius iQue Human ADCP Kit 5 x 96
Supplier:  Sartorius BA97107
Encompass
Perform high-throughput analysis: Validated reagents for rapid quantification of ADCP in 96 or 384-well platesObtain quantitative data: Generate reproducible and specific measurements of ADCP suitable for pharmacological analysisAnalyze multiple cell models: Assess ADCP on a variety of adherent or non-adherent target cellsMaximize productivity: Reduce time to answer with real time data analysis and novel visualization tools
Catalog No. 50-289-1614