Learn More
Sartorius iQue Human T Cell Activation
Supplier:  Sartorius 90562
Encompass
The iQue Human T Cell Activation Kit assesses the activation status of T cells while providing information about their health and their role in cytokine secretion. This one wash assay requires minimal hands on time and measures both cells and beads. Streamlined Workflow The iQue Human T Cell Activation Kit streamlines traditional workflow by measuring immune cell phenotypes early and late markers of T cell activation cell proliferation or encoded target cells cell viability and secreted cytokine concentrations IFNy and TNFa using only 5 μL-10 μL of samples. Analyze 15 endpoints from a single well assay. Simultaneous Measurement With the efficiency of the iQue Human T Cell Activation Kit you will be able to simultaneously measure T cell phenotypes activation markers cell proliferation cell viability and secreted cytokines in a single well.
Catalog No. 50-289-1650