Learn More
Sartorius iQue Membrane Integrity Kit
Catalog No. 502891604 Shop All Sartorius Products
Encompass
Three dye options gives you more ways to measure cell death.No wash protocol just add and read for fast easily automated workflow.Multiplex with other IntelliCyt kits like Cell Cycle for richer content.PrincipleThe Cell Membrane Integrity dyes are fluorescent molecules that are cell impermeant. Healthy cells with intact cell membranes will exclude the dyes. Unhealthy cells with compromised membranes will allow entry of the dye into the intracellular space. Upon entry to the cell the dye localizes to the nucleus and binds to DNA by intercalation.
Catalog No. 50-289-1604 Supplier Sartorius Supplier No. 90350
Three dye options gives you more ways to measure cell death.No wash protocol just add and read for fast easily automated workflow.Multiplex with other IntelliCyt kits like Cell Cycle for richer content.PrincipleThe Cell Membrane Integrity dyes are fluorescent molecules that are cell impermeant. Healthy cells with intact cell membranes will exclude the dyes. Unhealthy cells with compromised membranes will allow entry of the dye into the intracellular space. Upon entry to the cell the dye localizes to the nucleus and binds to DNA by intercalation.