Learn More
Sartorius iQue NK Cell Killing Kit
Catalog No. 502891736 Shop All Sartorius Products
Encompass
Speed: Sample to actionable data allows multiple assays to be run in one experiment thereby reducing the hands on time required numbers of overall tests and conserving precious sample.Results: Nine endpoints from a single well assay measuring activation markers CD69 and CD25 cell membrane integrity cell count secreted pro-inflammatory cytokine IFNγ and pro-apoptotic protease Granzyme B.Biological insights: Generate functional data and analyze cell health from same cell by multiplexing cell phenotypes and function in a single well.Ease of use: Single platform assay and data analysis with ForeCyt eliminates the time required for acquiring and correlating data from multiple sources.
Catalog No. 50-289-1736 Supplier Sartorius Supplier No. 97082
Speed: Sample to actionable data allows multiple assays to be run in one experiment thereby reducing the hands on time required numbers of overall tests and conserving precious sample.Results: Nine endpoints from a single well assay measuring activation markers CD69 and CD25 cell membrane integrity cell count secreted pro-inflammatory cytokine IFNγ and pro-apoptotic protease Granzyme B.Biological insights: Generate functional data and analyze cell health from same cell by multiplexing cell phenotypes and function in a single well.Ease of use: Single platform assay and data analysis with ForeCyt eliminates the time required for acquiring and correlating data from multiple sources.