Learn More
Sartorius iQue Proliferation&Encoding
Catalog No. 502891758 Shop All Sartorius Products
Encompass
Ability to encode cell and measure cell proliferation and using the violet laser.Multiplex with other MultiCyt kits like Cell Membrane Integrity or Qbeads for richer content.Dyes are safe and non-toxic to the cells.Covalently attaches to intracellular proteins enabling its long-term retention.PrincipleProliferation Use: The principle of dye dilution. Cell proliferation is detected and quantified based on the halving or “dilution” of the loaded dye after each round of cellular division. Through subsequent cell divisions daughter cells retain approximately half the fluorescent label of the parent cells.Encoding Use: Multiple cell populations are stained with different intensities of dye distinguish up to 4 cell populations Bright Mid Dim and Unstained . Target cells are stained with different concentrations of V/Blue encoding dye in order to discriminate the different cell populations.
Catalog No. 50-289-1758 Supplier Sartorius Supplier No. 97054
Ability to encode cell and measure cell proliferation and using the violet laser.Multiplex with other MultiCyt kits like Cell Membrane Integrity or Qbeads for richer content.Dyes are safe and non-toxic to the cells.Covalently attaches to intracellular proteins enabling its long-term retention.PrincipleProliferation Use: The principle of dye dilution. Cell proliferation is detected and quantified based on the halving or “dilution” of the loaded dye after each round of cellular division. Through subsequent cell divisions daughter cells retain approximately half the fluorescent label of the parent cells.Encoding Use: Multiple cell populations are stained with different intensities of dye distinguish up to 4 cell populations Bright Mid Dim and Unstained . Target cells are stained with different concentrations of V/Blue encoding dye in order to discriminate the different cell populations.