Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IR 783 98.0+%, TCI America™
SDP

Catalog No. I10311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10311G 1 g
I1031200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10311G Supplier TCI America Supplier No. I10311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 115970-66-6
Molecular Formula C38H46ClN2NaO6S2
Molecular Weight (g/mol) 749.354
MDL Number MFCD03093208
InChI Key QQIQAVJARACLHE-UHFFFAOYSA-M
Synonym 2-[2-[2-Chloro-3-[2-[1,3-dihydro-3,3-dimethyl-1-(4-sulfobutyl)-2H-indol-2-ylidene]ethylidene]-1-cyclohexen-1-yl]ethenyl]-3,3-dimethyl-1-(4-sulfobutyl)-3H-indolium Inner Salt Sodium Salt, Sodium 4-[2-[2-[2-Chloro-3-[2-[3,3-dimethyl-1-(4-sulfonatobutyl)indo
PubChem CID 71311115
IUPAC Name sodium;4-[(2Z)-2-[(2Z)-2-[2-chloro-3-[(E)-2-[3,3-dimethyl-1-(4-sulfonatobutyl)indol-1-ium-2-yl]ethenyl]cyclohex-2-en-1-ylidene]ethylidene]-3,3-dimethylindol-1-yl]butane-1-sulfonate
SMILES CC1(C2=CC=CC=C2[N+](=C1C=CC3=C(C(=CC=C4C(C5=CC=CC=C5N4CCCCS(=O)(=O)[O-])(C)C)CCC3)Cl)CCCCS(=O)(=O)[O-])C.[Na+]

Specifications

Melting Point 212°C
Quantity 1 g
Formula Weight 749.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material IR 783
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.