Learn More
Medchemexpress LLC IR-825 | 1558079-49-4 | 98.3% | 824.42 g/mol | C54H48ClN2O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY118924100MG
Encompass_Preferred
IR-825 is a near-infrared fluorescent dye with a terminal carboxyl (COOH) active group that enables covalent conjugation to biomolecules. It is used in imaging and photothermal therapy research due to its NIR absorbance and photothermal properties.
  • Near-infrared fluorescence for imaging and therapeutic studies.
  • Terminal carboxyl group enables bioconjugation.
  • High reported purity supports research applications.
  • Supplied in small laboratory package sizes for convenient handling.
Catalog No. 50-002-27537