Learn More
Medchemexpress LLC IR-825 near-infrared fluorescent dye | 1558079-49-4 | MFCD32068082 | 98.3% | 824.42 g/mol | C54H48ClN2O4 | 5 MG
Supplier: Medchemexpress LLC HY1189245MG
IR-825 is a near-infrared fluorescent dye with a terminal carboxyl (COOH) group that enables covalent conjugation to other molecules. It is used for near-infrared imaging and tumor photothermal research, and is supplied in research-scale quantities.
- Near-infrared fluorescent dye suitable for imaging
- Terminal carboxyl group enables conjugation to other molecules
- Used in tumor photothermal treatment research
- High purity (≈98.3%) for research applications
- Available in small research-scale packages (for example, 5 mg)
- Chemical formula C54H48ClN2O4; molecular weight 824.42 g/mol