Learn More
Strem, An Ascensus Company Ir(ppy)3, 95% 250mg
SDP
Supplier:  Strem, An Ascensus Company 777015250MG
Encompass
CAS# 94928-86-6. 250mg. Tris(2-phenylpyridinato-C2,N)iridium(III), 95%. . Molecular Weight: 654.78. Molecular Formula: C33H24IrN3. Color/form: yellow pwdr. Strem# 77-7015. http://www.strem.com/catalog/v/77-7015/
Catalog No. 50-203-4209