Learn More
Invitrogen™ IRAK4 Polyclonal Antibody

Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Horse and Bat.
Specifications
Specifications
| Antigen | IRAK4 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene | IRAK4 |
| Gene Accession No. | Q9NWZ3 |
| Gene Alias | 8430405M07Rik; 9330209D03Rik; interleukin 1 receptor associated kinase 4; interleukin-1; interleukin-1 receptor-associated kinase 4; IPD1; Irak4; IRAK-4; NY-REN-64; REN64; renal carcinoma antigen NY-REN-64 |
| Gene Symbols | IRAK4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.