Learn More
TARGETMOL CHEMICALS INC IRDABISANT 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T32191
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Irdabisant (CEP-26401) (CEP-26401) is a selective orally active and blood-brain barrier (BBB) penetrant antagonist/inverse agonist of histamine H3 receptor (H3R) (rat H3R Ki= 7.2 nM human H3R Ki= 2.0 nM). Irdabisant exhibits relatively low inhibitory activity against hERG current (IC50= 13.8 uM). Irdabisant exhibits cognition-enhancing and wake-promoting activities in the rat social recognition model. Irdabisant can be used for research on schizophrenia or cognitive impairment. purity: 99%
Catalog No. 50-258-0333
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.