Learn More
LI COR BIOTECH LLC IRDye 680RD Streptavidin (0.5 mg)
Supplier:  LI COR BIOTECH LLC 92668079
Encompass_Preferred
IRDye 680RD Streptavidin is supplied as a liquid in buffer containing 10 mM phosphate, 183 mM NaCl, 2.7 nM KCl, pH 7.4 with sodium azide 0.005% (w/v) as a preservative. To use, centrifuge briefly before use to eliminate aggregates that may have formed in solution. This will reduce non-specific background staining. A final concentration of 0.2 to 1.0 µg/ml (1:1,000 to 1:5,000) is usually satisfactory for most applications; however, appropriate dilution may need to be determined empirically. For membrane-based applications and In-Gel Westerns, it is recommended to add SDS (0.02% to 0.1% final concentration), in addition to Tween 20 (0.1 to 0.2% final concentration) during the detection incubation step to reduce non-specific background staining.
Catalog No. NC0337633
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.