Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRE1 alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110946 Supplier Invitrogen™ Supplier No. PA5110946
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTIGDKGECV ITPSTDVKFD PGLKSKNKLN YLRNYWLLIG HHETPLSAST KMLERFPNNL PKHRENVIPA DSEKKSFEEV INLVDQTSEN APTTVSRDVE EKPAHAPARP EAPVDSMLKD MATIILSTFL LIGWVA Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 90%.

ERN1 serine/threonine kinase or endoplasmic reticulum to nucleus signaling 1 is the ER to nucleus signaling 1 protein, a human homologue of the yeast Ire1 gene product which possesses intrinsic kinase activity and an endoribonuclease activity and it is important in endoplasmic reticulum-based stress signals. ERN1 controls IRE1 proteolysis in mammalian cells. ERN1 has a critical function in extraembryonic cells that is essential for fetal viability. Specifically, IRE1 acts as the sensor of unfolded proteins in the ER that initiates transmittance of the unfolded protein signal from the ER to the nucleus by splicing XBP1 mRNA converting it into a potent unfolded-protein response transcriptional activator. Null mutants are viable, but IRE1 is essential for viability under stress conditions that cause unfolded proteins to accumulate in the ER. Depletion of IRE1p through the expression of a dominant negative form of IRE1p has no effect on transfected cells, but cell death via apoptosis occurs under stress conditions that cause unfolded proteins to accumulate in the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRE1 alpha
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERN1
Gene Accession No. O75460
Gene Alias 9030414B18Rik; AI225830; C85377; endoplasmic reticulum (ER) to nucleus signalling 1; endoplasmic reticulum to nucleus signaling 1; endoplasmic reticulum to nucleus signalling 1; endoplasmic reticulum-to-nucleus signaling 1; Endoribonuclease; ER to nucleus signalling 1; ERN1; FLJ30999; hIRE1p; inositol-requiring 1; inositol-requiring 1 alpha; inositol-requiring enzyme 1; inositol-requiring protein 1; IRE1; IRE1a; IRE1a antibody; Ire1alpha; Ire1-alpha; Ire1p; IRE1p antibody; MGC163277; MGC163279; protein kinase/endoribonuclease; RGD1559716; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE1; serine/threonine-protein kinase/endoribonuclease IRE1-like
Gene Symbols ERN1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human IRE1 alpha. Recombinant protein control fragment (Product #RP-90501).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.