Learn More
Description
This antibody has 94% sequence homology with human, primate and rat. Suggested positive control: 3T3 whole cell lysate. Preliminary results have shown poor reactivity against human protein. Immunogen sequence homology: primate and rat (94%) proteins.
Specifications
Specifications
| Antigen | IREB2 |
| Applications | Gel Shift, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.15 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% sodium azide |
| Gene | IREB2 |
| Gene Accession No. | P48200, Q811J3 |
| Gene Alias | ACO3; D9Ertd85e; FLJ23381; im:7153062; Ireb2; IRE-BP 2; IREBP2; Iron regulatory protein 2; iron responsive element binding protein 2; iron-regulatory protein 2; iron-responsive element binding protein 2; iron-responsive element-binding protein 2; Iron-responsive element-binding protein 2-like protein; IRP2; IRP2AD |
| Gene Symbols | IREB2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
