Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IREM-2 (CD300e) Mouse anti-Human, Alexa Fluor 647, Clone: UP-H1, BD

Catalog No. BDB566315 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566315 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566315 Supplier BD Biosciences Supplier No. 566315
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The UP-H1 monoclonal antibody recognizes Immune receptor expressed by myeloid cells 2 (IREM-2) which is also known as CD300 antigen like family member E (CD300e), CMRF35-like molecule 2 (CLM2), or Polymeric immunoglobulin receptor 2 (PIgR2). IREM-2 is a 34 kDa type I transmembrane glycoprotein which belongs to the CMFRF35 subfamily of the immunoglobulin (Ig) superfamily. IREM-2 is expressed on monocytes, macrophages and myeloid dendritic cells (mDCs), but not on lymphoid dendritic cells or granulocytes. IREM-2 associates with DAP-12, an ITAM-containing adapter molecule, which provides activating signals. Crosslinking of IREM-2 leads to intracellular calcium mobilization, release of proinflammatory cytokines such as TNF, and increased expression of activation markers on monocytes and mDCs. IREM-2 activation of mDCs can reportedly enhance the alloreactive response of naive T cells.

Alexa Fluor™ 647 conjugates are highly photostable and remain fluorescent over a broad pH range. The excitation and emission maxima are nearly identical to those of APC. However, APC tends to be brighter while Alexa Fluor™ 647 is more optimal for intracellular applications. This fluorochrome exhibits uncommon photostability, making it an ideal choice for use in fluorescence microscopy. Due to nearly identical excitation and emission properties but different spillover characteristics, APC and Alexa Fluor™ 647 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IREM-2 (CD300e)
Classification Monoclonal
Clone UP-H1
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias IREM2, CD300E, CD300LE, CLM2, CLM-2, CMRF35-A5, PIgR-2, LMIR6
Host Species Mouse
Immunogen Human IREM-2 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.