Learn More
Signosis Inc IRF LUCIFERASE REPORTER HEK293
Supplier:  Signosis Inc SL0035
Encompass
IRF Responsive Luciferase Reporter HEK293 Cell Line is derived from human embryonic kidney, and stably express firefly luciferase reporter gene under the control of IRF response element. This cell line is an ideal cellular model for monitoring the activation of Immune Response Pathway triggered by stimuli treatment, enforced gene expression and gene knockdown
Catalog No. NC2337791
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.