Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRF7 Monoclonal Antibody (MNGPKL), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 501122335
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2335 100 μg
50-112-2334 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-2335 Supplier Invitrogen™ Supplier No. 12582982
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The monoclonal antibody MNGPKL reacts with mouse interferon regulatory factor 7 (IRF7), a 54 kDa transcription factor essential for induction of the antiviral innate immune response and IFN alpha production. B cells, plasmacytoid dendritic cells (pDC), and monocytes constitutively express IRF7 in the cytoplasm until activation by viral infection, double-stranded RNA or Toll-like receptor (TLR) signaling. CpG ODNs and single-stranded RNA, recognized by CD289 (TLR9) and TLR7, respectively, trigger IRF7 activation and upregulation. This activation promotes type I IFN response to disrupt pathogen and viral infection. These events cause IRF7 phosphorylation and translocation to the nucleus where it forms a homodimer to affect transcription. Applications Reported: This MNGPKL antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MNGPKL antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse splenocytes using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

Interferons (IFNs) are involved in a multitude of immune interactions during viral infections and play a major role in both the induction and regulation of innate and adaptive antiviral mechanisms. During infection, host-virus interactions signal downstream molecules such as transcription factors such as IFN regulatory factor-3 (IRF3) which can act to stimulate transcription of IFN-a/b genes. IRF7 has been shown to play a role in the transcriptional activation of virus-inducible cellular genes, including interferon beta chain genes. IRF7 play a major role in the innate immune pathway, interacting with the Toll-like receptor (TLR) adaptor proteins MyD88 and Tirp/TRAM and functioning as an intermediate TLR4 and TLR9 signaling. There are at least four differentially spliced isoforms of IRF7, although their function has not been clearly established.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF7
Applications Flow Cytometry
Classification Monoclonal
Clone MNGPKL
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene IRF7
Gene Accession No. P70434
Gene Alias IMD39; Interferon regulatory factor 7; interferon regulatory factor-7H; Irf7; IRF-7; IRF7A; IRF7B; IRF7C; IRF7H; IRF-7H
Gene Symbols IRF7
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54123
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.