Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRF7 Monoclonal Antibody (RDP4ND4), PE, eBioscience™

Catalog No. p-7174802
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-537-542 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-537-542 Supplier Invitrogen™ Supplier No. 12537542
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The monoclonal antibody RDP4ND4 recognizes human interferon regulatory factor 7 (IRF7), a 54 kDa member of the interferon regulatory transcription factor family. It is essential for the antiviral innate immune response and the activation of type I IFN genes. IRF7 is constitutively expressed in the cytoplasm of plasmacytoid dendritic cells, B cells and monocytes, but it is strongly induced in response to virus infection and interferon stimulation. IRF7 can activate IFN-beta and IFN-alpha genes via the virus-activated, MyD88-independent pathway, or the TLR-activated, MyD88-dependent pathway. Virus exposure induces IRF7, through toll-like receptors TLR7, TLR8, and TLR9, to promote IFN gene expression and disrupt infection. IRF7 exists in an inactive form until phosphorylated and its phosphorylation leads to dimerization and nuclear translocation of IRF7 where it aids in activating target genes. The IRF7 gene is upregulated by interferon stimulation through the Jak-STAT pathway in a positive feedback loop. Applications Reported: This RDP4ND4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RDP4ND4 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Please refer to BestProtocols®: Protocol B: One step protocol for (nuclear) intracellular proteins located under th...

Interferons (IFNs) are involved in a multitude of immune interactions during viral infections and play a major role in both the induction and regulation of innate and adaptive antiviral mechanisms. During infection, host-virus interactions signal downstream molecules such as transcription factors such as IFN regulatory factor-3 (IRF3) which can act to stimulate transcription of IFN-a/b genes. IRF7 has been shown to play a role in the transcriptional activation of virus-inducible cellular genes, including interferon beta chain genes. IRF7 play a major role in the innate immune pathway, interacting with the Toll-like receptor (TLR) adaptor proteins MyD88 and Tirp/TRAM and functioning as an intermediate TLR4 and TLR9 signaling. There are at least four differentially spliced isoforms of IRF7, although their function has not been clearly established.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF7
Applications Flow Cytometry
Classification Monoclonal
Clone RDP4ND4
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IRF7
Gene Accession No. Q92985
Gene Alias IMD39; Interferon regulatory factor 7; interferon regulatory factor-7H; Irf7; IRF-7; IRF7A; IRF7B; IRF7C; IRF7H; IRF-7H
Gene Symbols IRF7
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3665
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.