Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium(III) chloride hydrate, 99.8% (metals basis)

Catalog No. AA1103014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1103014 25 g
AA1103003 1 g
AA1103006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1103014 Supplier Thermo Scientific Chemicals Supplier No. 01103014
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14996-61-3 | Cl3Ir | 298.57 g/mol

Iridium(III) chloride hydrate is used to prepare other iridium compounds. It finds use as a catalyst for oxidation in presence of cerium(IV) and in the the manufacture of electrodes. Iridium(III) chloride is used in iridium electroplating, in making the standard meter bar of Paris and crucibles. It is used for high temperature electrical contacts, as hardening agent, in tipping pens and compass bearings and Pt/Ir spark plugs. It is used for medical, surgical or other patient oriented applications, MMO (mixed metal oxide) titanium electrode coatings and as a starting material for heterogeneous catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14996-61-3
Molecular Formula Cl3Ir
Molecular Weight (g/mol) 298.57
MDL Number MFCD00149750
InChI Key DANYXEHCMQHDNX-UHFFFAOYSA-K
IUPAC Name iridium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ir+3]

Specifications

Color Black
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Soluble in water and alcohol.
Formula Weight 298.56 (Anhydrous)
Odor Odorless
Chemical Name or Material Iridium(III) chloride hydrate
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 233-044-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.