Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium Standard, 1000μg/mL in water with dilute hydrochloric acid, Ultra Scientific

Catalog No. USICP077
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICP077 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICP077 Supplier Agilent Technologies Supplier No. ICP077
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7439-88-5 , 7647-01-0 , 7439-88-5 , 7732-18-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

CAS Max % 89.9, 10.0, 0.1
Analyte or Component Names Iridium,water,hydrochloric acid
Element Ir, Hydrochloric Acid
Solvent or Matrix Water/Dilute Hydrochloric Acid
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Iridium Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.