Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Irinotecan hydrochloride

Catalog No. AAJ60743MD
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60743MD 250 mg
AAJ60743MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60743MD Supplier Thermo Scientific Chemicals Supplier No. J60743MD
Add to Cart

Topoisomerase I inhibitor | CAS: 100286-90-6 | C33H39ClN4O6 | 623.15 g/mol

Topoisomerase I inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100286-90-6
Molecular Formula C33H39ClN4O6
Molecular Weight (g/mol) 623.15
MDL Number MFCD01862255
InChI Key GURKHSYORGJETM-WAQYZQTGSA-N
Synonym irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe
PubChem CID 74990
ChEBI CHEBI:5971
IUPAC Name hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride
SMILES [H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O

Specifications

Color Yellow
Quantity 250 mg
Merck Index 14,5091
Solubility Information Soluble in DMSO or DMF at approximately 20mg/ml; Sparingly soluble in aqueous buffers; /n
Formula Weight 623.14
Physical Form Powder
Chemical Name or Material Irinotecan hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber DW1061000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.