Learn More
Description
The IRIS9 Plus Prestained Protein Ladder is a combination of 9 pre-stained proteins with molecular weights from 15 to 170 kDa. The 9 recombinant proteins are covalently coupled with blue chromophore, while 1 green band at 15 kDa, a red band at 70 kDa and a newly designed peacock green band at 60 kDa serve as reference bands. The IRIS9 Plus Prestained Protein Ladder keeps track of the size and separation of proteins during SDS-polyacrylamide gel electrophoresis, approximating the target protein size and validating the Western transfer efficiency on PVDF, nylon, or nitrocellulose membranes.
Specifications
Specifications
| Formulation | 20 mM Trisphosphate at pH 7.5, 2% SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15% (v/v) Glycerol. |
| For Use With (Application) | 3ul or 5ul per loading for clear visualization during electrophoresis on 15-well or 10-well mini-gel, respectively. 2.5ul per well for general Western transferring. |
| Quantity | 500 μL |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
