Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron, 1000μg/mL, BAKER INSTRA-ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003120 Shop All Avantor Products
Change view
Click to view available options
Quantity:
150 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-003-120 150 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-120 Supplier Avantor Supplier No. 645304

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
IUPAC Name iron
SMILES [Fe]

Specifications

Packaging Narrow Mouth Poly Bottle
Specific Gravity 1L = 1.02kg
Quantity 150 mL
Formula Weight 55.85
Grade BAKER INSTRA-ANALYZED™ Reagent
Chemical Name or Material Iron, 1000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.