Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron, Aldon
SCI_ED

Catalog No. S96863 Shop All Aldon Products
Change view
Click to view available options
Metal Mesh (40-60 Mesh); 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S96863 Metal Mesh (40-60 Mesh); 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96863 Supplier Aldon Corporation Supplier No. IS18033

missing translation for 'validMainframeMsg'

Chemical Identifiers

Linear Formula Fe
Formula Weight 55.85
Chemical Name or Material Iron

Specifications

Color Gray to Black
Melting Point 2800
CAS 7439-89-6
Quantity 500 g
Packaging Metal Mesh (40-60 Mesh)
Molecular Formula Fe
MDL Number MFCD00010999
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES [Fe]
IUPAC Name iron
Molecular Weight (g/mol) 55.85
PubChem CID 23925
ChEBI CHEBI:82664
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife Indefinite
DOTInformation DOT Class 4.1, : Flammable Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.